missing translation for 'onlineSavingsMsg'
Learn More

Hesperetin 97.0+%, TCI America™

Catalog No. H07215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07215G 5 g
H072125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07215G Supplier TCI America Supplier No. H07215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 520-33-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00075646
InChI Key AIONOLUJZLIMTK-AWEZNQCLSA-N
Synonym hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1
PubChem CID 72281
ChEBI CHEBI:28230
IUPAC Name (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 232°C
Color White-Yellow
Quantity 5 g
Formula Weight 302.28
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hesperetin