missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HES5 Recombinant Rabbit Monoclonal Antibody (23GB6360)
GREENER_CHOICE

Numéro de catalogue. MA553026
Modifier l'affichage
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA553026 100 μL
MA553027 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553026 Fournisseur Invitrogen™ Code fournisseur MA553026
Il en reste null

Rabbit Recombinant Monoclonal Antibody

HES5 Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Hairy and Enhancer-of-split homologues 5 (HES5) isa helix-loop-helix transcriptional factor that negatively regulates cell differentiation during embryogenesis. It contains one basic helix-loop-helix (bHLH) domain and one Orange domain. HES5 is reported to be a down stream effector of the Notch Signaling Pathway and is expressed in advanced ovarian serous adenocarcinomas as well as in cervical squamous carcinoma. Reports also suggest that HES5 is abundantly expressed in osteoarthritis along with Notch1 and Jagged1.

Spécifications

Antigen HES5
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6360
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HES5
Gene Accession No. P70120, Q03062, Q5TA89
Gene Alias BHLHB38; Class B basic helix-loop-helix protein 38; hairy and enhancer of split 5; hairy and enhancer of split 5 (Drosophila); hes family bHLH transcription factor 5; Hes5; Hes-5; RP3-395M20.5; Transcription factor HES-5
Gene Symbols HES5
Host Species Rabbit
Immunogen A synthesized peptide derived from human HES5 (1-166AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15208, 388585, 79225
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats