Learn More
Alkali Scientific Hermle Z207-M High-Speed Bench
Numéro de catalogue. 503029681
Encompass
The Hermle Z207-M Compact Centrifuge machine is the perfect microcentrifuge for laboratories with valuable limited space who require quiet and powerful centrifugation The Hermle Z207-Ms compact design houses a powerful maintenance free induction motor that can reach up to 17317 xg Accompanied by an easy to use control panel the unique EZ-Scrolltouch pad gives you ease of control and a modern feel An advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10 In addition to retrieving saved programs the easy to use front panel can be used to adjust accel/decel rates run time and change the end of run alarm tone There is also a momentary spin function that will run while the button is pressed Additionally the Hermle Z207-M features a sleep mode to save energy by shutting off after a certain time if not being used while on which can be toggled on or off easily All common microtubes 0 2ml 0 5ml 1 5ml 2 0ml can be ut
Numéro de catalogue. 503029681 Fournisseur Alkali Scientific Code fournisseur RS6207M
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Hermle Z207-M Compact Centrifuge machine is the perfect microcentrifuge for laboratories with valuable limited space who require quiet and powerful centrifugation The Hermle Z207-Ms compact design houses a powerful maintenance free induction motor that can reach up to 17317 xg Accompanied by an easy to use control panel the unique EZ-Scrolltouch pad gives you ease of control and a modern feel An advanced microprocessor controls allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10 In addition to retrieving saved programs the easy to use front panel can be used to adjust accel/decel rates run time and change the end of run alarm tone There is also a momentary spin function that will run while the button is pressed Additionally the Hermle Z207-M features a sleep mode to save energy by shutting off after a certain time if not being used while on which can be toggled on or off easily All common microtubes 0 2ml 0 5ml 1 5ml 2 0ml can be ut