missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Heratherm Chambre de stabilité 750 L

Numéro de catalogue. 13642656
Modifier l'affichage
Click to view available options
Style de porte:
Glass door
Solid outer door with inner glass door
Solide (extérieur), Verre (intérieur)
Verre
Capacité (métrique):
247 L
750 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Style de porte Capacité (métrique)
13642656 Solid outer door with inner glass door 247 L
01672438 Solide (extérieur), Verre (intérieur) 750 L
01672439 Verre 750 L
13642655 Glass door 247 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 13642656 Fournisseur Thermo Scientific™ Code fournisseur ESS250

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Gagnez en confiance en votre produit lors des tests de stabilité et de durée de conservation avec des chambres de stabilité robustes Heratherm conçues pour répondre à diverses exigences de contrôle qualité.

Contrôles de température et d’humidité, bien adaptés aux tests de stabilité, à la durée de vie et aux tests environnementaux.

  • Broad relative humidity range to cover many applications, including humidity levels below ambient.
  • Environmentally friendly water-efficient humidification system.
  • Minimal condensate in standard applications.
  • Integrated water tank; fixed water connection for in-house purified water option available.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)

Spécifications

Types d’alarme Audible and Visible
Capacité (impériale) 8.8 cu.ft.
Capacité (impériale) d’étagère 66.1 lb.
Capacité (métrique) d’étagère 30 kg
Certifications/Conformité UL, CE
Sorties de données USB, Thermo Cloud via OPC UA
Capacité (métrique) 247 L
Matériau de la cuve Powder-coated Cold Rolled Steel
Éclairage No
Matériau Powder-coated Cold Rolled Steel
Nb d’étagères 2
Type de prise Nema 5-20P
Humidité relative 10 to 90%
Plage de températures (métrique) 0 to 70°C
Catégorie Stability Chamber
Affichage 7 in. Touchscreen user interface
Style de porte Solid outer door with inner glass door
À utiliser avec (application) Stability testing, environmental testing, storage
Plage d’humidité 10 to 90% RH
Source d’humidité Removable water tank
Nb de roulettes None
Ports 2 access ports
Programmabilité Temperature and humidity ramping
Zone des étagères (impériale) 3.05 ft.2
Superficie d’étagère (métrique) 0.28 m2
Matériau de l’étagère Stainless Steel (304/1.4301)
Contrôle de la température Microprocessor - programmable
Résolution de la température 0.1°C
Stabilité thermique ≤ ±0.1°C @ 20°C to 37°C
Uniformité de la température ≤ ±0.5°C @ 20°C to 37°C
Garantie 24 months
Consommation d’eau ≤ 0.25 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Résistivité de l’eau 0.05 - 1 MΩ
Largeur (impériale) 35.5 in.
Largeur (impériale) intérieure 26 in.
Largeur (impériale) d’étagère 24.2 in.
Largeur (métrique) 901 mm
Largeur (métrique) intérieure 660 mm
Largeur (métrique) d’étagère 615 mm
Hauteur (impériale) 44.4 in.
Hauteur (impériale) intérieure 31.5 in
Hauteur (métrique) 1128 mm
Hauteur (métrique) intérieure 800 mm
Profondeur (impériale) 31.1 in.
Profondeur (impériale) intérieure 20.4 in.
Profondeur (métrique) 790 mm
Profondeur (métrique) intérieure 519 mm
Poids à l’expédition (impériale) 459 lb.
Poids à l’expédition (métrique) 208 kg
Exigences électriques 100/240V, 50/60Hz
Tension 100/115/220/230 V
Puissance 900 W
Afficher plus de résultats Afficher moins de résultats