missing translation for 'onlineSavingsMsg'
Learn More

Heptanophenone 98.0+%, TCI America™

Numéro de catalogue. H036925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H036925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H036925G Fournisseur TCI America Code fournisseur H036925G
Il en reste null

Identifiants chimiques

CAS 1671-75-6
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00009539
InChI Key UXMQORVHJMUQFD-UHFFFAOYSA-N
Synonym heptanophenone, 1-heptanone, 1-phenyl, n-heptanophenone, unii-kx07wp06jy, kx07wp06jy, enanthophenone, hexyl phenyl ketone, 1-phenyl-heptan-1-one, 1-phenyl-1-heptanone #
PubChem CID 74282
IUPAC Name 1-phenylheptan-1-one
SMILES CCCCCCC(=O)C1=CC=CC=C1

Spécifications

Color Yellow
Boiling Point 283°C
Quantity 25 g
Formula Weight 190.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptanophenone