missing translation for 'onlineSavingsMsg'
Learn More

Heptanes, Technical, Reagents

Numéro de catalogue. 01337227
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
01337227 1 L
01337226 20 L
01337225 4 L
01337224 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 01337227 Fournisseur Labchem Inc Code fournisseur C22130001C
Il en reste null

Heptanes, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Identifiants chimiques

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym Heptanes (mixed isomers)
IUPAC Name heptane
SMILES CCCCCCC

Spécifications

Melting Point -90.6°C
Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.6795
Boiling Point 98.4°C
Grade Technical
Chemical Name or Material Heptanes, Technical