missing translation for 'onlineSavingsMsg'
Learn More

Heptane, Practical, Spectrum™ Chemical

Numéro de catalogue. p-7165629
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
18610790 500 mL Steel Can
18610789 4 L Steel Can
18610787 20 L Steel Can
18610788 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 18610790 Fournisseur Spectrum Chemical Mfg Cor Code fournisseur H1006500MLGL
Il en reste null

Identifiants chimiques

CAS 142-82-5
Molecular Weight (g/mol) 100.2

Spécifications

CAS Max % 1
Quantity 500 mL
Packaging Steel Can
Boiling Point 98.4°C
Grade Practical
Chemical Name or Material Heptane

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.