missing translation for 'onlineSavingsMsg'
Learn More

Heptane OmniSolv™, MilliporeSigma™

Catalog No. MHX00761
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX00761 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX00761 Supplier Technidata Supplier No. HX00761

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Low UV Cutoff

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91.11°C
Color Colorless
Density 0.68g/cm3 at 20°C
Quantity 1 L
Flash Point −4.15°C (−24.5°F), Closed cup
Ignition Point 221.85°C (431.3°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
UN Number 1206
Boiling Point 98.34°C
Formula Weight 100.2
Percent Purity ≥99.0%
Chemical Name or Material Heptane
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept uprightto prevent leakage.