missing translation for 'onlineSavingsMsg'
Learn More

Heptane, MilliporeSigma™

Catalog No. 50947601
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50947601 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947601 Supplier Technidata Supplier No. HX00903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point −90.5°C
Quantity 20 L
Flash Point −4°C
Packaging Steel Drum
Vapor Pressure 48 hPa (20°C)
Boiling Point 97°C to 98°C (1013 hPa)
Solubility Information 0.05 g/L
Recommended Storage Store at room temperature