missing translation for 'onlineSavingsMsg'
Learn More

Heptane 99.0+%, TCI America™

Numéro de catalogue. H0491500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H0491500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H0491500ML Fournisseur TCI America Code fournisseur H0491500ML
Il en reste null

Identifiants chimiques

Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Spécifications

Melting Point -91°C
Color Colorless
Quantity 500 mL
Physical Form Clear Liquid at 20°C
UN Number 1206
Boiling Point 98°C
Formula Weight 100.21
Percent Purity ≥99.0% (GC)
Chemical Name or Material Heptane [for HPLC Solvent]
EINECSNumber (2)-0007
RTECSNumber MI7700000
TSCA Yes