missing translation for 'onlineSavingsMsg'
Learn More

Heptane, MilliporeSigma™

Catalog No. MHX00781
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX00781 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX00781 Supplier Technidata Supplier No. US1HX0078/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, eptani
PubChem CID 8900
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Quantity 4 L
Packaging Glass Bottle
Linear Formula CH3(CH2)5CH3
Formula Weight 100.21
Chemical Name or Material Heptane