missing translation for 'onlineSavingsMsg'
Learn More

Heptane, MilliporeSigma™

Catalog No. MHX00801
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MHX00801 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MHX00801 Supplier Technidata Supplier No. HX00801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -90.5°C
Quantity 4 L
Flash Point -4−C
Packaging Glass Bottle
Physical Form Clear Liquid
Residue after Evaporation 5ppm max
Vapor Pressure 48 hPa (20°C)
Water 0.01% max
Boiling Point 97°C to 98°C (1013 hPa)
Formula Weight 100.21
Chemical Name or Material Heptane
Show More Show Less
EINECSNumber 205-563-8
RTECSNumber MI7700000
storageNote1 Store at room temperature.