missing translation for 'onlineSavingsMsg'
Learn More

Heptane, 99.0% min., MilliporeSigma™

Catalog No. MHX00806
Change view
Click to view available options
Quantity:
500 mL
200 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MHX00806 500 mL
MHX008020 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MHX00806 Supplier Technidata Supplier No. HX00806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Assay Percent Range 99.0% min.
Formula Weight 100.21
Chemical Name or Material Heptane