missing translation for 'onlineSavingsMsg'
Learn More

Heptakis(2,6-di-o-methyl)-β-cyclodextrin, MP Biomedicals™

Catalog No. ICN15732005
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15732005 5 g
ICN15732001 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15732005 Supplier MP Biomedicals Inc Supplier No. 0215732005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Heptakis(2,6-di-O-methyl)-β-cyclodextrin is a cholesterol sequestering agent

Chemical Identifiers

CAS 51166-71-3
Molecular Formula C56H98O35
Molecular Weight (g/mol) 1331.365
MDL Number MFCD00011616
InChI Key QGKBSGBYSPTPKJ-UZMKXNTCSA-N
Synonym 2,6-di-o-methyl-beta-cyclodextrin, unii-5fjp73e35o, dimethyl-beta-cyclodextrin, heptakis 2,6-o-dimethyl beta-cyclodextrin, heptakis-2,6-di-o-methyl-beta-cyclodextrin, heptakis 2,6-di-o-methyl-beta-cyclodextrin, dm-beta-cyd, dimethyl, a-cyclodextrin, 2,6-dme-beta-cyclodextrin, 2,6-di-o-methyl-beta-cd
PubChem CID 10171019
SMILES COCC1C2C(C(C(O1)OC3C(OC(C(C3O)OC)OC4C(OC(C(C4O)OC)OC5C(OC(C(C5O)OC)OC6C(OC(C(C6O)OC)OC7C(OC(C(C7O)OC)OC8C(OC(O2)C(C8O)OC)COC)COC)COC)COC)COC)COC)OC)O

Specifications

Color White
Quantity 5 g
Formula Weight 1331.4
Physical Form Powder
Chemical Name or Material Heptakis(2, 6-di-o-methyl)-β-cyclodextrin