missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptadecylbenzene 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 316.57 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Heptadecylbenzene |
Safety and Handling
| EINECSNumber | (3)-0021 |
| TSCA | Yes |