missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanoic acid, 98%

Catalog No. AAL0592806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0592806 5 g
AAL0592814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0592806 Supplier Thermo Scientific Chemicals Supplier No. L0592806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 506-12-7 | C17H34O2 | 270.457 g/mol

Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heptadecanoic acid is used as a reference during the analysis of medium-chain fatty acids from biomaterials such as seed oils and plant and animal oils.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly). Insoluble in water.

Notes
Store at 4°C. Store away from oxidizing agents.

Chemical Identifiers

CAS 506-12-7
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.457
MDL Number MFCD00002751
InChI Key KEMQGTRYUADPNZ-UHFFFAOYSA-N
Synonym margaric acid, n-heptadecanoic acid, margarinic acid, n-heptadecylic acid, n-heptadecoic acid, heptadecylic acid, fatty acids, c16-18, unii-v987y9oz8l, normal-heptadecanoic acid, margarinate
PubChem CID 10465
ChEBI CHEBI:32365
IUPAC Name heptadecanoic acid
SMILES CCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 59°C to 63°C
Density 0.853
Boiling Point 227°C (100 mmHg)
Linear Formula CH3(CH2)15CO2H
Quantity 5 g
Beilstein 1781004
Merck Index 14,5751
Solubility Information Soluble in Chloroform (Slightly),Methanol (Slightly). Insoluble in water.
Formula Weight 270.46
Percent Purity 98%
Chemical Name or Material Heptadecanoic acid
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 208-027-1
RTECSNumber MI3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only