missing translation for 'onlineSavingsMsg'
Learn More

Heptadecafluoro-n-octyl Iodide 98.0+%, TCI America™

Catalog No. P108425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P108425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P108425G Supplier TCI America Supplier No. P108425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 507-63-1
Molecular Formula C8F17I
Molecular Weight (g/mol) 545.965
MDL Number MFCD00001064
InChI Key KWXGJTSJUKTDQU-UHFFFAOYSA-N
Synonym perfluorooctyl iodide, heptadecafluoro-1-iodooctane, 1-iodoperfluorooctane, perfluoro-n-octyl iodide, 1-iodoheptadecafluorooctane, perfluoro-1-iodooctane, perfluorooctyliodide, heptadecafluoro-n-octyl iodide, octane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodo, octane, heptadecafluoro-1-iodo
PubChem CID 10491
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-heptadecafluoro-8-iodooctane
SMILES C(C(C(C(C(F)(F)I)(F)F)(F)F)(F)F)(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Color White-Yellow
Boiling Point 161°C
Quantity 25 g
Formula Weight 545.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Heptadecafluoro-n-octyl Iodide
EINECSNumber (2)-0090
RTECSNumber RG9700050
TSCA Yes