missing translation for 'onlineSavingsMsg'
Learn More

Heptacosane, ≥98% (GC), MilliporeSigma™ Supelco™

$119.97 - $406.00

Specifications

CAS 593-49-7
Flash Point 120°C
Physical Form Neat
Linear Formula CH3(CH2)25 CH3
MDL Number MFCD00009862
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111014796
View Documents
Merck Emd Millipore
51559250MG
250 mg
Each for $119.97
Only null left
 
111014795
View Documents
Merck Emd Millipore
515591G
1 g
Each for $406.00
Only null left
 
Description

Description

Featured Industries: Petroleum
Specifications

Specifications

593-49-7
Neat
MFCD00009862
270°C (15 mmHg)
380.73
Analytical Standard
120°C
CH3(CH2)25 CH3
58°C to 60°C
Analytical Standard
≥98% (GC)
Videos
SDS
Documents

Documents