missing translation for 'onlineSavingsMsg'
Learn More

Heptacosane, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014796
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014796 250 mg
111014795 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014796 Supplier Merck Emd Millipore Supplier No. 51559250MG
Featured Industries: Petroleum

Specifications

CAS 593-49-7
Flash Point 120°C
Physical Form Neat
Linear Formula CH3(CH2)25 CH3
MDL Number MFCD00009862
Melting Point 58°C to 60°C
Boiling Point 270°C (15 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 380.73
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less