missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heptacosane, ≥98% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 515591G
Description
Featured Industries: PetroleumSpecifications
| 593-49-7 | |
| Neat | |
| MFCD00009862 | |
| 270°C (15 mmHg) | |
| 1 g | |
| ≥98% (GC) |
| 120°C | |
| CH3(CH2)25 CH3 | |
| 58°C to 60°C | |
| Analytical Standard | |
| 380.73 | |
| Analytical Standard |