missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ HepG2 Whole Cell Lysate
Description
The HepG2 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation, SDS-Page.
Specifications
Specifications
| Quantity | 1 mg |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 4X Laemmli Sample Buffer [62.5mM Tris HCl, 1% LDS, 10% Glycerol, 0.005% bromophenol blue, pH 6.8] BME has not been added. |
| Test Specificity | HepG2 |
| Cellular Fraction | Whole |
| Host Species | Human |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| Protein State | Native |
| For Use With (Application) | Immunoprecipitation, SDS-Page, Western Blot |
| Tissue | Liver |
For Research Use Only