missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ HEPES, Sodium Salt, ULTROL™ Grade, Calbiochem™,
A zwitterionic N-substituted aminosulfonic acid buffer
$244.00 - $1180.00
Chemical Identifiers
| CAS | 75277-39-3 |
|---|---|
| Molecular Formula | C8H17N2NaO4S |
| Molecular Weight (g/mol) | 260.28 |
| MDL Number | MFCD00036463 |
| InChI Key | RDZTWEVXRGYCFV-UHFFFAOYSA-M |
| Synonym | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid, Na |
| PubChem CID | 2724248 |
| ChEBI | CHEBI:46758 |
| IUPAC Name | sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| SMILES | [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
391333100GM
|
MilliporeSigma™
391333100GM |
100 g |
Each for $244.00
|
|
|||||
|
391333500GM
|
MilliporeSigma™
391333500GM |
500 g |
Each for $887.00
|
|
|||||
|
3913331KG
|
MilliporeSigma™
3913331KG |
1 kg |
Each for $1,180.00
|
|
|||||
Description
HygroscopicChemical Identifiers
| 75277-39-3 | |
| 260.28 | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| 2724248 | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| C8H17N2NaO4S | |
| MFCD00036463 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid, Na | |
| CHEBI:46758 | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Specifications
| 75277-39-3 | |
| ≥99.0% | |
| MFCD00036463 | |
| Soluble in water | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
| 260.28 | |
| CHEBI:46758 | |
| by titration (dry basis) |
| 100 g | |
| C8H17N2NaO4S | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid, Na | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
| 2724248 | |
| 260.3 | |
| zwitterionic N-substituted aminosulfonic acid buffer |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.