missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HEPES sodium salt, ≥99%, MP Biomedicals™
HEPES is widely used in many biochemical reactions and as a buffering agent in some cell culture media. It is used in a protein deposition technique in electron microscopy because it did not affect metal substrates
Manufacturer: Mp Biomedicals 0219482890
Specifications
HEPES sodium salt | |
≥99% | |
MFCD00036463 | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate | |
2724248 | |
260.3 | |
White |
75277-39-3 | |
C8H17N2NaO4S | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt | |
C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+] | |
260.284 | |
CHEBI:46758 | |
Powder | |
500g |
Chemical Identifiers
75277-39-3 | |
260.284 | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
2724248 | |
sodium;2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonate |
C8H17N2NaO4S | |
MFCD00036463 | |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt | |
CHEBI:46758 | |
C1CN(CCN1CCO)CCS(=O)(=O)[O-].[Na+] |
Safety and Handling
EINECSNumber : 278-169-7