missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ HEPES, Free Acid, ULTROL™ Grade, Calbiochem™,
A zwitterionic N-substituted aminosulfonic acid buffer
$151.00 - $3890.00
Chemical Identifiers
| CAS | 7365-45-9 |
|---|---|
| Molecular Formula | C8H18N2O4S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00006158 |
| InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
| Synonym | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid |
| PubChem CID | 23831 |
| ChEBI | CHEBI:42334 |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
| SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
391338100GM
|
MilliporeSigma™
391338100GM |
100 g |
Each for $151.00
|
|
|||||
|
391338500GM
|
MilliporeSigma™
391338500GM |
500 g |
Each for $562.00
|
|
|||||
|
3913381KG
|
MilliporeSigma™
3913381KG |
1 kg |
Each for $825.00
|
|
|||||
|
3913385KG
|
MilliporeSigma™
3913385KG |
5 kg |
Each for $3,890.00
|
|
|||||
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 23831 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
| C8H18N2O4S | |
| MFCD00006158 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid | |
| CHEBI:42334 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
Specifications
| 7365-45-9 | |
| ≥99.0% | |
| MFCD00006158 | |
| Soluble in water | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 | |
| by titration (dry basis) |
| 100 g | |
| C8H18N2O4S | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 23831 | |
| 238.3 | |
| zwitterionic N-substituted aminosulfonic acid buffer |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area.