missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ HEPES, Free Acid, ULTROL™ Grade, Calbiochem™,
Specifications
Specifications
| pH | 5.0 to 6.5 |
| Physical Form | Crystalline Powder |
| Quantity | 5 kg |
| Solution Type | Buffer |
| Assay Percent Range | ≥99.0% |
| Solubility Information | Soluble in water |
| Formula Weight | 238.3 |
| Percent Purity | ≥99% (titration, dry basis) |
| Grade | by titration (dry basis) |
| Chemical Name or Material | zwitterionic N-substituted aminosulfonic acid buffer |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. |