missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HEPES, Free Acid, ULTROL™ Grade, 1.0 M Solution, Calbiochem™,

Catalog No. p-4953787
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
375368100ML 100 mL
375368500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 375368100ML Supplier MilliporeSigma™ Supplier No. 375368100ML
Only null left

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Quantity 100 mL
Assay Percent Range ≥99.0%
Solubility Information Not available
Formula Weight 238.3
Grade by titration (dry basis)
Concentration or Composition (by Analyte or Components) 1.0M solution
Chemical Name or Material HEPES, Free Acid
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).