missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ HEPES, Free Acid, OmniPur™, Calbiochem™,

Catalog No. 53405KG
Change view
Click to view available options
Quantity:
500 g
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53405KG 5 kg
5320500GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53405KG Supplier MilliporeSigma™ Supplier No. 53405KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Melting Point 210°C to 215°C
For Use With (Application) Molecular Biology
Quantity 5 kg
DNase None Detected
Assay Percent Range ≥99.0%
Protease None Detected
Solubility Information Soluble in water
Formula Weight 238.3
Percent Purity ≥99%
Grade Dried basis
Chemical Name or Material HEPES, Free Acid
Show More Show Less
Recommended Storage 15°C to 30°C (Do not freeze); Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use.