missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ HEPES, Free Acid, Molecular Biology Grade, Calbiochem™,
A zwitterionice N-substituted aminosulfonic acid buffer
$88.00 - $472.00
Chemical Identifiers
| CAS | 7365-45-9 |
|---|---|
| Molecular Formula | C8H18N2O4S |
| Molecular Weight (g/mol) | 238.30 |
| MDL Number | MFCD00006158 |
| InChI Key | JKMHFZQWWAIEOD-UHFFFAOYSA-N |
| Synonym | N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid |
| PubChem CID | 23831 |
| ChEBI | CHEBI:42334 |
| IUPAC Name | 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
| SMILES | OCCN1CCN(CCS(O)(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
39134025GM
|
MilliporeSigma™
39134025GM |
25 g |
Each for $88.00
|
|
|||||
|
391340250GM
|
MilliporeSigma™
391340250GM |
250 g |
Each for $472.00
|
|
|||||
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 23831 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid |
| C8H18N2O4S | |
| MFCD00006158 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid | |
| CHEBI:42334 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
Specifications
| 7365-45-9 | |
| None Detected | |
| C8H18N2O4S | |
| None Detected | |
| Soluble in water | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 | |
| by alkalimetric assay |
| 25 g | |
| ≥99.0% | |
| MFCD00006158 | |
| N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic Acid | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid | |
| 23831 | |
| 238.3 | |
| zwitterionic N-substituted aminosulfonic acid buffer |
Safety and Handling
Recommended Storage : 15°C to 30°C (Do not freeze).