missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ HEPES, Free Acid, Molecular Biology Grade, Calbiochem™,
Specifications
Specifications
| For Use With (Application) | Molecular Biology |
| pH | 6.8 to 8.2 |
| Physical Form | Solid |
| Quantity | 250 g |
| DNase | None Detected |
| Assay Percent Range | ≥99.0% |
| Protease | None Detected |
| Solubility Information | Soluble in water |
| Formula Weight | 238.3 |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
| Recommended Storage | 15°C to 30°C (Do not freeze). |