missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES free acid, 99+%, for molecular biology, DNase, Rnase and Protease free

Catalog No. AC329850500
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC329850500 50 g
AC329855000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC329850500 Supplier Thermo Scientific Chemicals Supplier No. 329850500

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Absorbance (0.5 M in water), (0.5 M in water) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
Absorbance Conditions (0.5M in water)
Melting Point 234.0°C to 238.0°C
Color White
pH 5 to 6.5 (0.5 M aq. soln.)
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99+%
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (105°C)
Packaging Plastic bottle
Beilstein 23, V,2, 379
Merck Index 15, 4689
Solubility Information Solubility in water: 704g/L (20°C).
Formula Weight 238.3
Percent Purity ≥99%
Grade Molecular Biology
Chemical Name or Material HEPES free acid
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 230-907-9
RTECSNumber TL6809000
TSCA TSCA

RUO – Research Use Only