missing translation for 'onlineSavingsMsg'
Learn More

HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Catalog No. BP3101
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP3101 1 kg Poly Bottle
BP310100 100 g Glass Bottle
BP310500 500 g Poly Bottle
BP3105 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP3101 Supplier Fisher BioReagents Supplier No. FLBP310 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Commonly used buffering agent | CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Commonly used buffering agent

HEPES is commonly used as a buffering agent.

Specifications

Chemical Name or Material HEPES
Absorbance 0.01 max. (0.1M solution) at 280nm
CAS 7365-45-9
Color White
pH 5.0 to 6.5
Physical Form Powder or Solid
Quantity 1 kg
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Assay Percent Range ≥99 %
Identification Pass Test
Molecular Formula C8H18N2O4S
MDL Number MFCD00006158
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Solubility Information Soluble in water
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Weight (g/mol) 238.30
PubChem CID 23831
ChEBI CHEBI:42334
Percent Purity ≥99%
Grade Molecular Biology
Show More Show Less