missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES-buffered saline, pH 7.0 (5X)

Catalog No. p-7041931
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60753AK 250 mL
AAJ60753AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60753AK Supplier Thermo Scientific Chemicals Supplier No. J60753AK
Only null left

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES is widely used in cell culture. HEPES is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization. N-2-hydroxyethylpiperazine-N-2?-ethanesulfonic acid (Hepes) has been tested as an alternative to tris-(hydroxymethyl)aminoethane (Tris) for hydrogen-ion buffering of algal cultures. Separation of human platelets from plasma proteins including factor VIII VWF by a combined albumin gradient-gel filtration method by using HEPES buffer. May be used in isoelectric focusing applications and as a buffer in the quantitative and selective measurement of antigen-antibody reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is widely used in cell culture. HEPES is used to neutralize the complex proteins in growth medium that may inactivate trypsin during trypsinization. N-2-hydroxyethylpiperazine-N-2′-ethanesulfonic acid (Hepes) has been tested as an alternative to tris-(hydroxymethyl)aminoethane (Tris) for hydrogen-ion buffering of algal cultures. Separation of human platelets from plasma proteins including factor VIII VWF by a combined albumin gradient-gel filtration method by using HEPES buffer. May be used in isoelectric focusing applications and as a buffer in the quantitative and selective measurement of antigen-antibody reactions.

Solubility
Soluble in H2O: 1 M at 20°C, clear, colorless

Notes
Contains 125mM HEPES and 750mM sodium chloride, at pH 7.0

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 5X
pH 7.0
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Solubility Information Soluble in H2O: 1 M at 20°C,clear,colorless
Chemical Name or Material HEPES-buffered saline
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only