missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES-buffered saline, pH 6.5 (5X)

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Supplier:  Thermo Scientific Chemicals J61239AP

 View more versions of this product

Catalog No. AAJ61239AP


Only null left

Description

Description

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility
Fully miscible in water.

Notes
Contains 125mM HEPES and 750mM sodium chloride, pH 6.5
Specifications

Specifications

HEPES-buffered saline
Colorless
6.5
500 mL
Odorless
Fully miscible in water.
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
7365-45-9
5X
Liquid
C8H18N2O4S
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only