missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES-buffered saline, pH 6.5 (5X)

Catalog No. AAJ61239AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61239AK 250 mL
AAJ61239AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61239AK Supplier Thermo Scientific Chemicals Supplier No. J61239AK

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility
Fully miscible in water.

Notes
Contains 125mM HEPES and 750mM sodium chloride, pH 6.5

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color Colorless
Concentration 5X
pH 6.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Solubility Information Fully miscible in water.
Chemical Name or Material HEPES-buffered saline
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only