missing translation for 'onlineSavingsMsg'
Learn More

HEPES Buffer, 1M Solution, Cell Culture Grade, MP Biomedicals

Catalog No. ICN1688449
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN1688449 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN1688449 Supplier MP Biomedicals Inc Supplier No. 091688449

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

1M solution pH 7.2 to 7.4 at 37°C Storage temperature: 15-30°C

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1 M
For Use With (Application) Mammalian Cell Culture
Physical Form Solution
Quantity 100 mL
Solution Type Buffer
Grade Cell Culture
Chemical Name or Material Hepes Buffer