missing translation for 'onlineSavingsMsg'
Learn More

HEPES, ≥99% (Dry Basis), Ultrapure

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Supplier:  Thermo Fisher Scientific J1692630

 View more versions of this product

Catalog No. AAJ1692630


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

HEPES
7365-45-9
250 g
C8H18N2O4S
Plastic bottle
N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
≥99%
∼236°C (decomposition)
Crystalline Powder
Dry basis
MFCD00006158
883043
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
238.3

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C8H18N2O4S
MFCD00006158
N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P312-P501c

H335

EINECSNumber : 230-907-9

RTECSNumber : TL6809000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only