missing translation for 'onlineSavingsMsg'
Learn More

HEPES, ≥99% (Dry Basis), Ultrapure

Catalog No. AAJ16926A7
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ16926A7 5 kg Plastic bottle
AAJ1692622 100 g Plastic bottle
AAJ1692630 250 g Plastic bottle
AAJ16926A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ16926A7 Supplier Thermo Fisher Scientific Supplier No. J16926A7

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym N-[2-Hydroxyethyl]piperazine-N'-[2-Ethanesulfonic acid]
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Melting Point ∼236°C (decomposition)
Physical Form Crystalline Powder
Quantity 5 kg
Packaging Plastic bottle
Beilstein 883043
Formula Weight 238.3
Percent Purity ≥99%
Assay Dry basis
Chemical Name or Material HEPES
Hazard Category H335
Precautionary Statement P261-P271-P304+P340-P312-P501c
EINECSNumber 230-907-9
RTECSNumber TL6809000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only