missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 99%, for biochemistry

Catalog No. p-4510574
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC172571000 100 g Glass bottle
AC172572500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC172571000 Supplier Thermo Scientific Chemicals Supplier No. 172571000
Only null left

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
Synonym 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Absorbance 0.08 max. at 260nm, 0.05 max. at 280nm, 0.08 max. at 250nm, (5% in water) 1 cm cell, (5% in water) 1 cm cell at 0nm
Absorbance Conditions (5% aq. soln.) (1cm cell)
Melting Point 234.0°C to 238.0°C
Color (5% in water) clear, colorless
Decomposition Information 238°C
Physical Form Liquid or Solid
Quantity 100 g
Assay Percent Range 99%
Infrared Spectrum Authentic
Loss on Drying 1 % max. (1 g, 105°C, 3 hrs)
Packaging Glass bottle
Beilstein 23, V,2, 379
Merck Index 15, 4689
Solubility Information Solubility in water: 704g/L (20°C).
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Formula Weight 238.3
Percent Purity 99%
Grade Biochemical
Chemical Name or Material HEPES
Show More Show Less

RUO – Research Use Only