missing translation for 'onlineSavingsMsg'
Learn More

HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure

Catalog No. p-7082831
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ16924AE 100 mL Nalgene bottle
AAJ16924AP 500 mL Nalgene bottle
AAJ16924K2 1 L Nalgene bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ16924AE Supplier Thermo Fisher Scientific Supplier No. J16924AE
Only null left

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1 M
pH 7.3
Physical Form Solution
Quantity 100 mL
Packaging Nalgene bottle
Beilstein 883043
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Grade Molecular Biology
Chemical Name or Material HEPES
EINECSNumber 230-907-9
RTECSNumber TL6809000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only