missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 8.5

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

Supplier:  Thermo Scientific Chemicals J61360AK

 View more versions of this product

Catalog No. AAJ61360AK


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HEPES buffer soln.
1.0 M
Liquid
C8H18N2O4S
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
23831
7365-45-9
8.5
250 mL
MFCD00006158
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
CHEBI:42334
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
23831
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only