missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 7.0

Catalog No. p-7042482
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62688AK 250 mL
AAJ62688AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ62688AK Supplier Thermo Scientific Chemicals Supplier No. J62688AK
Only null left

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH. It acts a buffering component, which is used in the preparation of buffers. It is described as one of the best all-purpose buffers available for use in biological research.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.  

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Color Colorless
Concentration 1.0 M
pH 7.0
Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with water.
Chemical Name or Material HEPES buffer soln.
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only