missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES, 1.0M buffer soln., pH 6.5
CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Supplier: Thermo Scientific Chemicals J61017AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| HEPES buffer soln. | |
| 1.0 M | |
| Liquid | |
| C8H18N2O4S | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
| 23831 |
| 7365-45-9 | |
| 6.5 | |
| 500 mL | |
| MFCD00006158 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 | |
| 238.30 | |
| CHEBI:42334 |
Chemical Identifiers
| 7365-45-9 | |
| 238.30 | |
| JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
| CHEBI:42334 | |
| OCCN1CCN(CCS(O)(=O)=O)CC1 |
| C8H18N2O4S | |
| MFCD00006158 | |
| 23831 | |
| 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only