missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals HEPES, 0.5M buffer soln., pH 8.0

Catalog No. AAJ63002AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63002AE 100 mL
AAJ63002AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63002AE Supplier Thermo Scientific Chemicals Supplier No. J63002AE

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose Good’s buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose “Good’s” buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 0.5 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Solubility Information Fully miscible.
Chemical Name or Material HEPES buffer soln.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only