missing translation for 'onlineSavingsMsg'
Learn More

Air-Tite Products™ Henke-Ject Low Dead Space Syringe, 1 mL

Catalog No. 14910159
Change view
Click to view available options
Volume (Metric):
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14910159 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14910159 Supplier Air-Tite Products™ Supplier No. RL1
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Henke-Ject Low Dead Space Syringe offers a low dead space plunger and clean gliding motion for a smooth draw.

Henke-Ject Low Dead Space syringes offer a low dead space plunger and smooth gliding motion. These high quality syringes have a lubricated, synthetic rubber tipped piston to allow for smooth draw. A positive stop prevents accidental pullout. These syringes are ETO gas sterilized and are latex- and pyrogen-free.

  • 3 part syringe
  • Smooth, gliding motion
  • Silicone lubricant
  • Works with any ISO needle

Specifications

Certifications/Compliance ISO 13485
Material Polypropylene
Latex Free Yes
Disposable Single-use
Graduations 0.2 mL
Quantity 100 Pk.
Volume (Metric) 1 mL
For Use With (Equipment) Hypodermic Needle
Connection Type Luer Lock
Graduated Yes
Sterility Sterile
Show More Show Less