missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Hemoglobin alpha Polyclonal Antibody
GREENER_CHOICE

Numéro de catalogue. PIPA579347
Modifier l'affichage
Click to view available options
Quantité:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIPA579347 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIPA579347 Fournisseur Invitrogen™ Code fournisseur PA579347

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue. IHC: human placenta tissue, human tonsil tissue. Flow: K562 cell.

The human alpha globin gene cluster located on chromosome 16 spans about 30 kb and includes seven loci: 5'- zeta - pseudozeta - mu - pseudoalpha-1 - alpha-2 - alpha-1 - theta - 3'. The alpha-2 (HBA2) and alpha-1 (HBA1) coding sequences are identical. These genes differ slightly over the 5' untranslated regions and the introns, but they differ significantly over the 3' untranslated regions. Two alpha chains plus two beta chains constitute HbA, which in normal adult life comprises about 97% of the total hemoglobin; alpha chains combine with delta chains to constitute HbA-2, which with HbF (fetal hemoglobin) makes up the remaining 3% of adult hemoglobin. Alpha thalassemias result from deletions of each of the alpha genes as well as deletions of both HBA2 and HBA1; some nondeletion alpha thalassemias have also been reported.

Spécifications

Antigène Hemoglobin alpha
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugué Unconjugated
Formule PBS with 4mg trehalose and no preservative
Gène Hba1
Numéro d’acquisition du gène P69905
Alias du gène alpha 1 globin; alpha globin 1; alpha one globin; alpha-1/2-globin; alpha-2 globin chain; alpha-globin; delta globin; GloA; globin, alpha; haemaglobin alpha 1; Hba; HBA1; HBA2; Hba-a1; Hba-a2; HBAM; Hbat1; HBA-T3; HBH; hemoglobin alpha 1 chain; hemoglobin alpha 1 globin chain; hemoglobin alpha chain; hemoglobin alpha, adult chain 1; hemoglobin alpha, adult chain 2; hemoglobin alpha-1/2 chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin subunit alpha-1/2; hemoglobin, alpha 1; LOC287167
Symboles de gène Hba1
Espèces hôtes Rabbit
Immunogène E.coli-derived human Hemoglobin recombinant protein (Position: V2-R142).
Méthode de purification Antigen affinity chromatography
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 3039
Espèces cibles Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Lyophilized
Isotype IgG
Afficher plus de résultats Afficher moins de résultats