Learn More
HemoCue America WBC Microcuvettes
Catalog No. 22601018
Encompass
Change view
Click to view available options
Capacity (Metric):
10 μL
Catalog No. Capacity (Metric)
22601018 10 μL
1 options
Catalog No. 22601018 Supplier HemoCue America Supplier No. 039113003
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Serves as a sample container and reaction chamber
  • Made of plastic
  • Comprises a body having a cavity that holds about 5 or 10μL of sample
  • Cavity contains reagents deposited on its inner walls
  • Blood sample is drawn into the cavity by capillary action and mixes with the reagents
Specifications
Type Micro Cuvette
Capacity (Metric) 10 μL
Material Plastic
For Use With (Equipment) HemoCue WBC system
For Use With (Application) Serves as a sample container and reaction chamber