missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ HeLa Whole Cell Lysate
Description
The HeLa Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Quantity | 0.1 mg |
| Storage Buffer | Standard RIPA buffer including protease inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. Denaturing agents have not been added. |
| Purity | Protein |
| Cellular Fraction | Whole |
| Content And Storage | Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. |
| Product Line | Novus |
| For Use With (Application) | Western Blot |
| Tissue | Reproductive |
For Research Use Only