Learn More
Heidolph™ Jacketed Process Reaction Vessel Kits
Radleys jacketed process vessel kits for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
$9,124.15 - $26,658.74
Specifications
For Use With (Equipment) Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors
Includes Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
Max. Temperature (English) 446°F
View More Specs
Includes: Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
Products 12
Catalog Number Mfr. No. Volume (Metric) Vessel Type Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Vessel Type Price Quantity & Availability  
13890003
View Documents Promotion Details
Heidolph™
015952350
100 mL Single Jacketed
Each for $9,124.15
Only null left
13890004
View Documents Promotion Details
Heidolph™
015950114
100 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890005
View Documents Promotion Details
Heidolph™
015952360
250 mL Single Jacketed
Each for $9,940.63
Only null left
13890007
View Documents Promotion Details
Heidolph™
015951120
500 mL Single Jacketed
Each for $9,913.19
Only null left
13890006
View Documents Promotion Details
Heidolph™
015951115
250 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890008
View Documents Promotion Details
Heidolph™
015950189
500 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890010
View Documents Promotion Details
Heidolph™
015952170
1000 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890009
View Documents Promotion Details
Heidolph™
015950187
1000 mL Single Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890011
View Documents Promotion Details
Heidolph™
015950188
2000 mL Single Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890012
View Documents Promotion Details
Heidolph™
015952200
2000 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890014
View Documents Promotion Details
Heidolph™
015950190
5000 mL Vacuum Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
13890013
View Documents Promotion Details
Heidolph™
015952060
5000 mL Single Jacketed N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • These cost effective vessel kits include the vessel and accessories you need to get started.
  • The Pt100 probe and turbine stirrer included in each kit are matched to the vessel size.
  • Each vessel kit includes reaction vessel, PTFE turbine stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
  • Process Vessels - mimic the geometry of plant scale reactors
  • Process vessels feature a 1.25 to 1 ratio of internal height to internal diameter, with dish-shaped bottom
Specifications
Specifications
Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors
Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
446°F