Learn More
Heidolph™ Jacketed Process Reaction Vessel Kits
Radleys jacketed process vessel kits for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
Supplier:  Heidolph™ 015952170
 View more versions of this product
Catalog No. 13890010
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes: Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
Description
Description
  • These cost effective vessel kits include the vessel and accessories you need to get started.
  • The Pt100 probe and turbine stirrer included in each kit are matched to the vessel size.
  • Each vessel kit includes reaction vessel, PTFE turbine stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
  • Process Vessels - mimic the geometry of plant scale reactors
  • Process vessels feature a 1.25 to 1 ratio of internal height to internal diameter, with dish-shaped bottom
Specifications
Chemical Identifiers
Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors
Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
446°F
Specifications
1000 mL
Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
Vacuum Jacketed
Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors
446°F
Product Suggestions
Product Suggestions