Learn More
Heidolph™ Jacketed Process Reaction Vessel Kits
Radleys jacketed process vessel kits for use with Reactor-Ready™ and Reactor-Ready™ Duo lab reactors
Supplier: Heidolph™ 015951120
Includes: Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter
Description
- These cost effective vessel kits include the vessel and accessories you need to get started.
- The Pt100 probe and turbine stirrer included in each kit are matched to the vessel size.
- Each vessel kit includes reaction vessel, PTFE turbine stirrer, side arm couplings, Pt100 PTFE temperature probe with LEMO connector, and PTFE temperature probe adapter.
- Process Vessels - mimic the geometry of plant scale reactors
- Process vessels feature a 1.25 to 1 ratio of internal height to internal diameter, with dish-shaped bottom
Chemical Identifiers
| Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors | |
| Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter | |
| 446°F |
Specifications
| 500 mL | |
| Process vessel, turbine stirrer paddle, PTFE Pt100 temp probe (w/Lemo plug for connection to the thermoregulator) and PTFE Temperature probe adapter | |
| Single Jacketed |
| Reactor-Ready™ and Reactor-Ready&Trade; Duo Lab Reactors | |
| 446°F |